missing translation for 'onlineSavingsMsg'
Learn More

Isopropylamine 99.0+%, TCI America™

Catalog No. I016525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I016525ML 25 mL
I0165500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I016525ML Supplier TCI America Supplier No. I016525ML

Chemical Identifiers

CAS 75-31-0
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.112
MDL Number MFCD00008082
InChI Key JJWLVOIRVHMVIS-UHFFFAOYSA-N
Synonym isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino
PubChem CID 6363
ChEBI CHEBI:15739
IUPAC Name propan-2-amine
SMILES CC(C)N

Specifications

Melting Point -95°C
Color Colorless
Boiling Point 34°C
Quantity 25 mL
UN Number 1221
Formula Weight 59.11
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Isopropylamine
EINECSNumber (2)-0131
RTECSNumber NT8400000
TSCA Yes