missing translation for 'onlineSavingsMsg'
Learn More

Isopropylamine Hydrochloride 98.0+%, TCI America™

Catalog No. I016625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I016625G 25 g
I0166100G 100 g
I0166500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I016625G Supplier TCI America Supplier No. I016625G
Only null left

Chemical Identifiers

CAS 15572-56-2
Molecular Formula C3H10ClN
Molecular Weight (g/mol) 95.57
MDL Number MFCD00050705
InChI Key ISYORFGKSZLPNW-UHFFFAOYSA-N
Synonym 2-Aminopropane Hydrochloride, Isopropylammonium Chloride
PubChem CID 6432137
IUPAC Name propan-2-amine;hydrochloride
SMILES CC(C)N.Cl

Specifications

Melting Point 160°C
Color White
Quantity 25 g
Formula Weight 95.57
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropylamine Hydrochloride
EINECSNumber (2)-0131
RTECSNumber NT9530000
TSCA Yes