missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Trifluoroacetate 98.0+%, TCI America™

Catalog No. T176125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T176125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T176125G Supplier TCI America Supplier No. T176125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 400-38-4
Molecular Formula C5H7F3O2
Molecular Weight (g/mol) 156.10
MDL Number MFCD00013566
InChI Key ASAXRKSDVDALDT-UHFFFAOYSA-N
Synonym isopropyl trifluoroacetate, isopropyltrifluoroacetate, isopropyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, 1-methylethyl ester, iso-propyl trifluoroacetate, trifluoroacetic acid isopropyl ester, pubchem12590, acmc-1ctsh, methylethyl 2,2,2-trifluoroacetate
PubChem CID 78998
IUPAC Name propan-2-yl 2,2,2-trifluoroacetate
SMILES CC(C)OC(=O)C(F)(F)F

Specifications

Color Colorless
Boiling Point 73°C
Quantity 25 g
UN Number 2924
Formula Weight 156.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Trifluoroacetate