missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl trifluoroacetate, 97%

Catalog No. p-7023599
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1732214 25 g
AAA1732222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1732214 Supplier Thermo Scientific Chemicals Supplier No. A1732214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 400-38-4 | C5H7F3O2 | 156.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 400-38-4
Molecular Formula C5H7F3O2
Molecular Weight (g/mol) 156.10
MDL Number MFCD00013566
InChI Key ASAXRKSDVDALDT-UHFFFAOYSA-N
Synonym isopropyl trifluoroacetate, isopropyltrifluoroacetate, isopropyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, 1-methylethyl ester, iso-propyl trifluoroacetate, trifluoroacetic acid isopropyl ester, pubchem12590, acmc-1ctsh, methylethyl 2,2,2-trifluoroacetate
PubChem CID 78998
IUPAC Name propan-2-yl 2,2,2-trifluoroacetate
SMILES CC(C)OC(=O)C(F)(F)F

Specifications

Melting Point <-78°C
Density 1.108
Boiling Point 73°C
Flash Point −8°C (17°F)
Refractive Index 1.319
Quantity 25 g
UN Number UN2924
Beilstein 1764312
Sensitivity Moisture sensitive
Formula Weight 156.1
Percent Purity 97%
Chemical Name or Material Isopropyl trifluoroacetate
Show More Show Less
Hazard Category H225-H315-H318-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 206-922-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only