missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Phosphate (Mono- and Di- Ester mixture), TCI America™
$65.24 - $244.19
Chemical Identifiers
| CAS | 52933-00-3 |
|---|---|
| Molecular Formula | C9H24O8P2 |
| Molecular Weight (g/mol) | 322.23 |
| MDL Number | MFCD00135965 |
| InChI Key | RTIXFJOJNSXSOB-UHFFFAOYSA-N |
| Synonym | phosphoric acid isopropyl ester, isopropyl phosphate mono-and di-ester mixture |
| PubChem CID | 44630356 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I024725ML
|
TCI America
I024725ML |
25 mL |
Each for $65.24
|
|
|||||
|
I0247500ML
|
TCI America
I0247500ML |
500 mL |
Each for $244.19
|
|
|||||
Chemical Identifiers
| 52933-00-3 | |
| 322.23 | |
| RTIXFJOJNSXSOB-UHFFFAOYSA-N | |
| 44630356 |
| C9H24O8P2 | |
| MFCD00135965 | |
| phosphoric acid isopropyl ester, isopropyl phosphate mono-and di-ester mixture |
Specifications
| 52933-00-3 | |
| C9H24O8P2 | |
| 25 mL | |
| phosphoric acid isopropyl ester, isopropyl phosphate mono-and di-ester mixture | |
| 322.23 | |
| 182.16 | |
| Isopropyl Phosphate (Mono- and Di- Ester mixture) |
| Yellow | |
| MFCD00135965 | |
| 1793 | |
| RTIXFJOJNSXSOB-UHFFFAOYSA-N | |
| 44630356 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1981&(2)-1986
RTECSNumber : TC6320000
TSCA : No