missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Myristate 98.0+%, TCI America™

Catalog No. p-7132194
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
M048125ML 25 mL
M0481500ML 500 mL
2 options

Catalog No. M048125ML

Supplier: TCI America M048125ML

Only null left

Chemical Identifiers

CAS 110-27-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00008982
InChI Key AXISYYRBXTVTFY-UHFFFAOYSA-N
Synonym isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester
PubChem CID 8042
IUPAC Name propan-2-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC(C)C

Specifications

Color Yellow
Boiling Point 193°C
Quantity 25 mL
Formula Weight 270.46
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Myristate
EINECSNumber (2)-0798
RTECSNumber XB8600000
TSCA Yes