missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Methyl Sulfide 97.0+%, TCI America™

Catalog No. I048325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I048325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I048325G Supplier TCI America Supplier No. I048325G
Only null left

Chemical Identifiers

CAS 1551-21-9
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.184
MDL Number MFCD00015035
InChI Key ROSSIHMZZJOVOU-UHFFFAOYSA-N
PubChem CID 15246
IUPAC Name 2-methylsulfanylpropane
SMILES CC(C)SC

Specifications

Color Yellow
Boiling Point 85°C
Quantity 25 g
UN Number 1993
Formula Weight 90.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Methyl Sulfide
TSCA No
Recommended Storage Refrigerator