missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Methanesulfonate 98.0+%, TCI America™
$74.56 - $188.27
Chemical Identifiers
| CAS | 926-06-7 |
|---|---|
| Molecular Formula | C4H10O3S |
| Molecular Weight (g/mol) | 138.181 |
| MDL Number | MFCD00047802 |
| InChI Key | SWWHCQCMVCPLEQ-UHFFFAOYSA-N |
| Synonym | isopropyl methanesulfonate, isopropyl mesylate, isopropylmethanesulfonate, methanesulfonic acid, 1-methylethyl ester, isopropyl methane sulphonate, 2-propyl methanesulphonate, isopropyl methane sulfonate, methanesulfonic acid, isopropyl ester, isopropyl methanesulphonate, unii-t0k2txy26b |
| PubChem CID | 13551 |
| IUPAC Name | propan-2-yl methanesulfonate |
| SMILES | CC(C)OS(=O)(=O)C |
Chemical Identifiers
| 926-06-7 | |
| 138.181 | |
| SWWHCQCMVCPLEQ-UHFFFAOYSA-N | |
| 13551 | |
| CC(C)OS(=O)(=O)C |
| C4H10O3S | |
| MFCD00047802 | |
| isopropyl methanesulfonate, isopropyl mesylate, isopropylmethanesulfonate, methanesulfonic acid, 1-methylethyl ester, isopropyl methane sulphonate, 2-propyl methanesulphonate, isopropyl methane sulfonate, methanesulfonic acid, isopropyl ester, isopropyl methanesulphonate, unii-t0k2txy26b | |
| propan-2-yl methanesulfonate |
Specifications
| 926-06-7 | |
| Yellow | |
| C4H10O3S | |
| 1 g | |
| isopropyl methanesulfonate, isopropyl mesylate, isopropylmethanesulfonate, methanesulfonic acid, 1-methylethyl ester, isopropyl methane sulphonate, 2-propyl methanesulphonate, isopropyl methane sulfonate, methanesulfonic acid, isopropyl ester, isopropyl methanesulphonate, unii-t0k2txy26b | |
| CC(C)OS(=O)(=O)C | |
| 138.181 | |
| 138.18 | |
| Liquid |
| 7°C | |
| 67°C | |
| MFCD00047802 | |
| 2810 | |
| SWWHCQCMVCPLEQ-UHFFFAOYSA-N | |
| propan-2-yl methanesulfonate | |
| 13551 | |
| ≥98.0% (GC) | |
| Isopropyl Methanesulfonate |
Safety and Handling
RTECSNumber : PB2275000
TSCA : Yes