missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Methanesulfonate 98.0+%, TCI America™

Catalog No. I09141G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09141G 1 g
I09145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09141G Supplier TCI America Supplier No. I09141G
Only null left

Chemical Identifiers

CAS 926-06-7
Molecular Formula C4H10O3S
Molecular Weight (g/mol) 138.181
MDL Number MFCD00047802
InChI Key SWWHCQCMVCPLEQ-UHFFFAOYSA-N
Synonym isopropyl methanesulfonate, isopropyl mesylate, isopropylmethanesulfonate, methanesulfonic acid, 1-methylethyl ester, isopropyl methane sulphonate, 2-propyl methanesulphonate, isopropyl methane sulfonate, methanesulfonic acid, isopropyl ester, isopropyl methanesulphonate, unii-t0k2txy26b
PubChem CID 13551
IUPAC Name propan-2-yl methanesulfonate
SMILES CC(C)OS(=O)(=O)C

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 67°C
Quantity 1 g
UN Number 2810
Formula Weight 138.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Methanesulfonate
RTECSNumber PB2275000
TSCA Yes