Learn More
Isopropyl methanesulfonate, 99%
CAS: 926-06-7 | C4H10O3S | 138.19 g/mol
Supplier: Thermo Scientific Chemicals 412840250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Isopropyl methanesulfonate | |
| 926-06-7 | |
| 1.1450g/mL | |
| 104°C | |
| 98.5% min. (GC) | |
| C4H10O3S | |
| CH3SO3CH(CH3)2 | |
| 25 g | |
| 1.145 | |
| SWWHCQCMVCPLEQ-UHFFFAOYSA-N | |
| propan-2-yl methanesulfonate | |
| 13551 | |
| 99% |
| 99% | |
| Colorless | |
| 82.0°C (6.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4180 to 1.4210 | |
| MFCD00047802 | |
| 04,IV,13 | |
| isopropyl methanesulfonate, isopropyl mesylate, isopropylmethanesulfonate, methanesulfonic acid, 1-methylethyl ester, isopropyl methane sulphonate, 2-propyl methanesulphonate, isopropyl methane sulfonate, methanesulfonic acid, isopropyl ester, isopropyl methanesulphonate, unii-t0k2txy26b | |
| CC(C)OS(=O)(=O)C | |
| 138.19 | |
| 138.19 | |
| Liquid |
Chemical Identifiers
| 926-06-7 | |
| 138.19 | |
| SWWHCQCMVCPLEQ-UHFFFAOYSA-N | |
| 13551 | |
| CC(C)OS(=O)(=O)C |
| C4H10O3S | |
| MFCD00047802 | |
| isopropyl methanesulfonate, isopropyl mesylate, isopropylmethanesulfonate, methanesulfonic acid, 1-methylethyl ester, isopropyl methane sulphonate, 2-propyl methanesulphonate, isopropyl methane sulfonate, methanesulfonic acid, isopropyl ester, isopropyl methanesulphonate, unii-t0k2txy26b | |
| propan-2-yl methanesulfonate |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Suspected of causing cancer.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 213-132-0
RTECSNumber : PB2275000
TSCA : TSCA
RUO – Research Use Only