missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Isobutyrate 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 120°C |
| Quantity | 25 mL |
| UN Number | 2406 |
| Formula Weight | 130.19 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Isopropyl Isobutyrate |
Safety and Handling
| EINECSNumber | (2)-0779 |
| RTECSNumber | NQ5320000 |
| TSCA | No |