missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Ether, Stabilized, ACS, 99%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 68°C (154.4°F) |
| Packaging | Amber Glass Bottle |
| Quantity | 250 mL |
| Residue after Evaporation | 0.0001 |
| Percent Purity | 0.99 |
| Grade | ACS |
| Titratable Acid | 0.0007 meq/g |
| Physical Form | Liquid |
| Chemical Name or Material | Isopropyl Ether, Stabilized |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.