missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-β-D-Thiogalactopyranoside, MP Biomedicals

Catalog No. ICN19402990
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19402990 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19402990 Supplier MP Biomedicals Inc Supplier No. 0219402990

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Molecular Biology Reagent. Cell culture media component for use in molecular genetics.β-Galactoside inducer

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside
PubChem CID 656894
ChEBI CHEBI:61448
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 500 mg
Formula Weight 238.3
Percent Purity ≥99%
Grade Not Available
Physical Form Powder
Chemical Name or Material Isopropyl-β-D-Thiogalactopyranoside
Recommended Storage 0°C