missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl-β-D-Thiogalactopyranoside, >99%, MP Biomedicals
Molecular Biology Reagent. Cell culture media component for use in molecular genetics.β-Galactoside inducer
Supplier: MP Biomedicals Inc 0219402991
| Quantity | 1 g |
|---|
Chemical Identifiers
| C9H18O5S | |
| MFCD00063273 | |
| iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside | |
| CHEBI:61448 |
Specifications
| Isopropyl-β-D-Thiogalactopyranoside | |
| >99% | |
| MFCD00063273 | |
| iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside | |
| CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O | |
| 238.30 | |
| CHEBI:61448 |
| 367-93-1 | |
| C9H18O5S | |
| 1 g | |
| BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol | |
| 656894 | |
| 238.3 |
Safety and Handling
Recommended Storage : 0°C