missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-β-D-Thiogalactopyranoside, >99%, MP Biomedicals

Catalog No. ICN19402980
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
10 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19402980 100 mg
ICN19402983 250 mg
ICN19402991 1 g
ICN19402994 5 g
ICN19402995 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN19402980 Supplier MP Biomedicals Inc Supplier No. 0219402980

Molecular Biology Reagent. Cell culture media component for use in molecular genetics.β-Galactoside inducer

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside
PubChem CID 656894
ChEBI CHEBI:61448
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Assay Percent Range >99%
Quantity 100 mg
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol
Formula Weight 238.3
Chemical Name or Material Isopropyl-β-D-Thiogalactopyranoside
Recommended Storage 0°C