missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Crotonate 98.0+%, TCI America™

Catalog No. p-7124842
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C118325ML 25 mL
C1183500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C118325ML Supplier TCI America Supplier No. C118325ML

Chemical Identifiers

CAS 6284-46-4
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00059053
InChI Key AABBHSMFGKYLKE-SNAWJCMRSA-N
Synonym Crotonic Acid Isopropyl Ester
PubChem CID 5354359
IUPAC Name propan-2-yl (E)-but-2-enoate
SMILES CC=CC(=O)OC(C)C

Specifications

Color Colorless
Boiling Point 147°C
Quantity 25 mL
UN Number 3272
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Crotonate
EINECSNumber (2)-0966
TSCA No