missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Chloroformate (ca. 30% in Toluene, ca. 2mol/L), TCI America™
$76.43 - $236.73
Chemical Identifiers
| CAS | 108-23-6 |
|---|---|
| Molecular Formula | C4H7ClO2 |
| Molecular Weight (g/mol) | 122.548 |
| MDL Number | MFCD00075068 |
| InChI Key | IVRIRQXJSNCSPQ-UHFFFAOYSA-N |
| Synonym | isopropyl chloroformate, isopropyl chlorocarbonate, carbonochloridic acid, 1-methylethyl ester, isopropyl chloromethanoate, isopropyl carbonochloridate, chloroformic acid isopropyl ester, unii-8lg1g18vxr, formic acid, chloro-, isopropyl ester, carbonochloride acid, 1-methylethyl ester, isopropylester kyseliny chlormravenci |
| PubChem CID | 7917 |
| IUPAC Name | propan-2-yl carbonochloridate |
| SMILES | CC(C)OC(=O)Cl |
Chemical Identifiers
| 108-23-6 | |
| 122.548 | |
| IVRIRQXJSNCSPQ-UHFFFAOYSA-N | |
| 7917 | |
| CC(C)OC(=O)Cl |
| C4H7ClO2 | |
| MFCD00075068 | |
| isopropyl chloroformate, isopropyl chlorocarbonate, carbonochloridic acid, 1-methylethyl ester, isopropyl chloromethanoate, isopropyl carbonochloridate, chloroformic acid isopropyl ester, unii-8lg1g18vxr, formic acid, chloro-, isopropyl ester, carbonochloride acid, 1-methylethyl ester, isopropylester kyseliny chlormravenci | |
| propan-2-yl carbonochloridate |
Specifications
| 108-23-6 | |
| C4H7ClO2 | |
| 100 g | |
| isopropyl chloroformate, isopropyl chlorocarbonate, carbonochloridic acid, 1-methylethyl ester, isopropyl chloromethanoate, isopropyl carbonochloridate, chloroformic acid isopropyl ester, unii-8lg1g18vxr, formic acid, chloro-, isopropyl ester, carbonochloride acid, 1-methylethyl ester, isopropylester kyseliny chlormravenci | |
| CC(C)OC(=O)Cl | |
| 122.548 | |
| 122.55 | |
| Isopropyl Chloroformate (ca. 30% in Toluene, ca. 2mol/L) |
| Colorless | |
| MFCD00075068 | |
| 3286 | |
| IVRIRQXJSNCSPQ-UHFFFAOYSA-N | |
| propan-2-yl carbonochloridate | |
| 7917 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1144&(2)-1704
RTECSNumber : LQ6475000
TSCA : Yes
Recommended Storage : Refrigerator