missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl bromoacetate, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.3990g/mL |
| Boiling Point | 59.0°C to 61.0°C (10.0 mmHg) |
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass Bottle |
| Refractive Index | 1.4440 to 1.4460 |
| Linear Formula | BrCH2CO2CH(CH3)2 |
| Quantity | 100 g |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. |
| EINECSNumber | 249-956-2 |
| TSCA | TSCA |
RUO – Research Use Only