missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Acetoacetate 97.0+%, TCI America™
$70.84 - $874.24
Chemical Identifiers
| CAS | 542-08-5 |
|---|---|
| Molecular Formula | C7H12O3 |
| Molecular Weight (g/mol) | 144.17 |
| MDL Number | MFCD00043933 |
| InChI Key | GVIIRWAJDFKJMJ-UHFFFAOYSA-N |
| Synonym | isopropyl acetoacetate, isopropyl 3-oxobutanoate, acetoacetic acid isopropyl ester, butanoic acid, 3-oxo-, 1-methylethyl ester, unii-38gtb2c6xc, 38gtb2c6xc, methylethyl 3-oxobutanoate, acip, isopropyl acetylacetate, isopropyl 3-oxobutyrate |
| PubChem CID | 68329 |
| IUPAC Name | propan-2-yl 3-oxobutanoate |
| SMILES | CC(C)OC(=O)CC(=O)C |
Chemical Identifiers
| 542-08-5 | |
| 144.17 | |
| GVIIRWAJDFKJMJ-UHFFFAOYSA-N | |
| 68329 | |
| CC(C)OC(=O)CC(=O)C |
| C7H12O3 | |
| MFCD00043933 | |
| isopropyl acetoacetate, isopropyl 3-oxobutanoate, acetoacetic acid isopropyl ester, butanoic acid, 3-oxo-, 1-methylethyl ester, unii-38gtb2c6xc, 38gtb2c6xc, methylethyl 3-oxobutanoate, acip, isopropyl acetylacetate, isopropyl 3-oxobutyrate | |
| propan-2-yl 3-oxobutanoate |
Specifications
| 542-08-5 | |
| 187°C | |
| MFCD00043933 | |
| isopropyl acetoacetate, isopropyl 3-oxobutanoate, acetoacetic acid isopropyl ester, butanoic acid, 3-oxo-, 1-methylethyl ester, unii-38gtb2c6xc, 38gtb2c6xc, methylethyl 3-oxobutanoate, acip, isopropyl acetylacetate, isopropyl 3-oxobutyrate | |
| CC(C)OC(=O)CC(=O)C | |
| 144.17 | |
| 144.17 | |
| Liquid |
| Yellow | |
| C7H12O3 | |
| 25 g | |
| GVIIRWAJDFKJMJ-UHFFFAOYSA-N | |
| propan-2-yl 3-oxobutanoate | |
| 68329 | |
| ≥97.0% (GC) | |
| Isopropyl Acetoacetate |
Safety and Handling
EINECSNumber : (2)-1475
TSCA : Yes