missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Acetoacetate 97.0+%, TCI America™

Catalog No. I0826500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0826500G 500 g
I082625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0826500G Supplier TCI America Supplier No. I0826500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 542-08-5
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00043933
InChI Key GVIIRWAJDFKJMJ-UHFFFAOYSA-N
Synonym isopropyl acetoacetate, isopropyl 3-oxobutanoate, acetoacetic acid isopropyl ester, butanoic acid, 3-oxo-, 1-methylethyl ester, unii-38gtb2c6xc, 38gtb2c6xc, methylethyl 3-oxobutanoate, acip, isopropyl acetylacetate, isopropyl 3-oxobutyrate
PubChem CID 68329
IUPAC Name propan-2-yl 3-oxobutanoate
SMILES CC(C)OC(=O)CC(=O)C

Specifications

Color Yellow
Boiling Point 187°C
Quantity 500 g
Formula Weight 144.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Acetoacetate
EINECSNumber (2)-1475
TSCA Yes