missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl 1-Thio-beta-D-galactopyranoside 98.0+%, TCI America™
$72.15 - $284.79
Chemical Identifiers
| CAS | 367-93-1 |
|---|---|
| Molecular Formula | C9H18O5S |
| Molecular Weight (g/mol) | 238.30 |
| MDL Number | MFCD00063273 |
| InChI Key | BPHPUYQFMNQIOC-NXRLNHOXSA-N |
| Synonym | iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside |
| PubChem CID | 656894 |
| ChEBI | CHEBI:61448 |
| IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol |
| SMILES | CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
Chemical Identifiers
| 367-93-1 | |
| 238.30 | |
| BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
| 656894 | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol |
| C9H18O5S | |
| MFCD00063273 | |
| iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside | |
| CHEBI:61448 | |
| CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
Specifications
| 367-93-1 | |
| White-Yellow | |
| MFCD00063273 | |
| BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol | |
| 656894 | |
| 238.30 | |
| Crystalline Powder |
| 113°C | |
| C9H18O5S | |
| iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside | |
| CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O | |
| 238.30 | |
| CHEBI:61448 | |
| ≥98.0% (GC) | |
| Isopropyl 1-Thio-beta-D-galactopyranoside |
Safety and Handling
TSCA : Yes