missing translation for 'onlineSavingsMsg'
Learn More

Isopropenyloxytrimethylsilane 90.0+%, TCI America™

Catalog No. I03245ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03245ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I03245ML Supplier TCI America Supplier No. I03245ML
Only null left

Chemical Identifiers

CAS 1833-53-0
Molecular Formula C6H14OSi
Molecular Weight (g/mol) 130.262
MDL Number MFCD00042845
InChI Key UAIFZYSPVVBOPN-UHFFFAOYSA-N
Synonym Trimethylisopropenyloxysilane
PubChem CID 74599
IUPAC Name trimethyl(prop-1-en-2-yloxy)silane
SMILES CC(=C)O[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 mL
UN Number 1993
Formula Weight 130.26
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropenyloxytrimethylsilane [Trimethylsilylating Agent]
EINECSNumber (2)-2960
TSCA Yes
Recommended Storage Refrigerator