missing translation for 'onlineSavingsMsg'
Learn More

Isopropenyl Acetate 98.0+%, TCI America™

Catalog No. p-7116674
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
A003525ML 25 mL
A0035500ML 500 mL
2 options

Catalog No. A003525ML

Supplier: TCI America A003525ML

Only null left

Chemical Identifiers

CAS 108-22-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008709
InChI Key HETCEOQFVDFGSY-UHFFFAOYSA-N
Synonym isopropenyl acetate, 2-acetoxypropene, 1-methylvinyl acetate, 2-acetoxypropylene, methylvinyl acetate, 1-propen-2-ol, acetate, propen-2-yl acetate, acetic acid isopropenyl ester, 1-acetoxy-1-methylethylene, 1-propen-2-yl acetate
PubChem CID 7916
IUPAC Name prop-1-en-2-yl acetate
SMILES CC(=C)OC(C)=O

Specifications

Melting Point -93°C
Color Yellow
Boiling Point 97°C
Quantity 25 mL
UN Number 2403
Formula Weight 100.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropenyl Acetate
EINECSNumber (2)-0729
RTECSNumber UD4200000
TSCA Yes