missing translation for 'onlineSavingsMsg'
Learn More

Isopropenyl Acetate 98.0+%, TCI America™

Catalog No. A003525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A003525ML 25 mL
A0035500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A003525ML Supplier TCI America Supplier No. A003525ML

Chemical Identifiers

CAS 108-22-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008709
InChI Key HETCEOQFVDFGSY-UHFFFAOYSA-N
Synonym isopropenyl acetate, 2-acetoxypropene, 1-methylvinyl acetate, 2-acetoxypropylene, methylvinyl acetate, 1-propen-2-ol, acetate, propen-2-yl acetate, acetic acid isopropenyl ester, 1-acetoxy-1-methylethylene, 1-propen-2-yl acetate
PubChem CID 7916
IUPAC Name prop-1-en-2-yl acetate
SMILES CC(=C)OC(C)=O

Specifications

Melting Point -93°C
Color Yellow
Boiling Point 97°C
Quantity 25 mL
UN Number 2403
Formula Weight 100.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropenyl Acetate
EINECSNumber (2)-0729
RTECSNumber UD4200000
TSCA Yes