missing translation for 'onlineSavingsMsg'
Learn More

Isoprene (stabilized with TBC) 99.0+%, TCI America™

Catalog No. p-7131244
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I016025ML 25 mL
I0160500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I016025ML Supplier TCI America Supplier No. I016025ML
Only null left

Chemical Identifiers

CAS 78-79-5
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00008600
InChI Key RRHGJUQNOFWUDK-UHFFFAOYSA-N
Synonym isoprene, 2-methyl-1,3-butadiene, isopentadiene, 2-methylbutadiene, 1,3-butadiene, 2-methyl, 2-methyldivinyl, beta-methylbivinyl, isopreno, isoterpene, isopren
PubChem CID 6557
ChEBI CHEBI:35194
IUPAC Name 2-methylbuta-1,3-diene
SMILES CC(=C)C=C

Specifications

Melting Point -146°C
Color Yellow
Boiling Point 35°C
Quantity 25 mL
UN Number 1218
Formula Weight 68.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoprene (stabilized with TBC)
EINECSNumber (2)-0020
RTECSNumber NT4037000
TSCA Yes
Recommended Storage Refrigerator