missing translation for 'onlineSavingsMsg'
Learn More

Isoprene monoxide, 95%

Catalog No. p-7058518
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1976806 5 g
AAL1976814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1976806 Supplier Thermo Scientific Chemicals Supplier No. L1976806
Only null left

CAS: 1838-94-4 | C5H8O | 84.118 g/mol

Isoprene monoxide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoprene monoxide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Recommended storage temperature is 2 - 8°C.

Chemical Identifiers

CAS 1838-94-4
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00075225
InChI Key FVCDMHWSPLRYAB-UHFFFAOYSA-N
Synonym 2-methyl-2-vinyloxirane, isoprene monoxide, ccris 5685, 3,4-epoxy-3-methyl-1-butene, 2-methyl-2-vinyl-oxirane, oxirane, 2-ethenyl-2-methyl-, 2r, acmc-20m5m5, oxirane, 2-ethenyl-2-methyl, 4,4,4-3fmet-1-2-furyl-1,3-butandione
PubChem CID 92166
IUPAC Name 2-ethenyl-2-methyloxirane
SMILES CC1(CO1)C=C

Specifications

Density 0.857
Boiling Point 79°C to 81°C
Flash Point −8°C (17°F)
Odor Recognizable
Refractive Index 1.416
Quantity 5 g
UN Number UN1993
Beilstein 104216
Solubility Information Slightly soluble in water.
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material Isoprene monoxide
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only