missing translation for 'onlineSavingsMsg'
Learn More

(-)-Isopinocampheylborane TMEDA Complex 98.0+%, TCI America™

Catalog No. i07975g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I07975G Supplier TCI America Supplier No. I07975G

Chemical Identifiers

CAS 67826-92-0
Molecular Formula C26H50B2N2
Molecular Weight (g/mol) 412.32
MDL Number MFCD00077801
InChI Key YIPGSNPACSKQKJ-NCHGFZPDSA-N
Synonym (-)-N,N′C-Bis(monoisopinocampheylborane)-N,N,N′C,N′C-tetramethylethylenediamine
PubChem CID 16211704
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine;[(1S,3S,4R,5S)-4,6,6-trimethyl-3-bicyclo[3.1.1]heptanyl]boron
SMILES [B]C1CC2CC(C1C)C2(C)C.[B]C1CC2CC(C1C)C2(C)C.CN(C)CCN(C)C

Specifications

Melting Point 141°C
Color White
Quantity 5 g
UN Number 1325
Formula Weight 416.35
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Isopinocampheylborane TMEDA Complex