missing translation for 'onlineSavingsMsg'
Learn More

Isopimpinellin 98.0+%, TCI America™

Catalog No. I086110MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I086110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I086110MG Supplier TCI America Supplier No. I086110MG
Only null left

Chemical Identifiers

CAS 482-27-9
Molecular Formula C13H10O5
Molecular Weight (g/mol) 246.22
MDL Number MFCD00017407
InChI Key DFMAXQKDIGCMTL-UHFFFAOYSA-N
Synonym 4,9-Dimethoxyfuro[3,2-g]benzopyran-7-one, 5,8-Dimethoxypsoralen
PubChem CID 68079
ChEBI CHEBI:28853
IUPAC Name 4,9-dimethoxy-7H-furo[3,2-g]chromen-7-one
SMILES COC1=C2OC=CC2=C(OC)C2=C1OC(=O)C=C2

Specifications

Melting Point 150°C
Quantity 10 mg
Formula Weight 246.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isopimpinellin
RTECSNumber LV1049200
TSCA No