missing translation for 'onlineSavingsMsg'
Learn More

Isophytol 95.0+%, TCI America™

Catalog No. I0145100ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0145100ML 100 mL
I014525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0145100ML Supplier TCI America Supplier No. I0145100ML
Only null left

Chemical Identifiers

CAS 505-32-8
Molecular Formula C20H40O
Molecular Weight (g/mol) 296.539
MDL Number MFCD00048380
InChI Key KEVYVLWNCKMXJX-UHFFFAOYSA-N
Synonym isophytol, 3,7,11,15-tetramethyl-1-hexadecen-3-ol, 1-hexadecen-3-ol, 3,7,11,15-tetramethyl, 2,6,10,14-tetramethylhexadec-15-en-14-ol, 2,6,10-trimethyl-14-vinylpentadecan-14-ol, 1-hexadecene-3-ol, 3,7,11,15-tetramethyl, 1-phyten-3-ol, iso-phytol, dsstox_cid_5474
PubChem CID 10453
IUPAC Name 3,7,11,15-tetramethylhexadec-1-en-3-ol
SMILES CC(C)CCCC(C)CCCC(C)CCCC(C)(C=C)O

Specifications

Color Yellow
Boiling Point 185°C
Quantity 100 mL
UN Number 3082
Formula Weight 296.54
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Isophytol
EINECSNumber (2)-0258
TSCA Yes