missing translation for 'onlineSavingsMsg'
Learn More

Isophytol, 95%

Catalog No. AAA1858318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1858318 50 g
AAA1858309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1858318 Supplier Thermo Scientific Chemicals Supplier No. A1858318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 505-32-8 | C20H40O | 296.539 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 505-32-8
Molecular Formula C20H40O
Molecular Weight (g/mol) 296.539
MDL Number MFCD00048380
InChI Key KEVYVLWNCKMXJX-UHFFFAOYSA-N
Synonym isophytol, 3,7,11,15-tetramethyl-1-hexadecen-3-ol, 1-hexadecen-3-ol, 3,7,11,15-tetramethyl, 2,6,10,14-tetramethylhexadec-15-en-14-ol, 2,6,10-trimethyl-14-vinylpentadecan-14-ol, 1-hexadecene-3-ol, 3,7,11,15-tetramethyl, 1-phyten-3-ol, iso-phytol, dsstox_cid_5474
PubChem CID 10453
IUPAC Name 3,7,11,15-tetramethylhexadec-1-en-3-ol
SMILES CC(C)CCCC(C)CCCC(C)CCCC(C)(C=C)O

Specifications

Density 0.8519
Boiling Point 125°C to 128°C (0.06 mmHg)
Refractive Index 1.4571
Quantity 50 g
Beilstein 1783243
Merck Index 14,5198
Formula Weight 296.54
Percent Purity 95%
Chemical Name or Material Isophytol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-008-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only