missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Isophthaloyl chloride
Description
Isophthaloyl chloride
Specifications
Specifications
| CAS | 99-63-8 |
| Melting Point | 43°C to 44°C |
| Boiling Point | 276°C |
| Molecular Formula | C8H4Cl2O2 |
| Linear Formula | C6H4-1,3-(COCl)2 |
| MDL Number | MFCD00000678 |
| Quantity | 500 g |
| Synonym | Isophthaloyl dichloride |
| Molecular Weight (g/mol) | 203.02 |
| Percent Purity | ≥99% |