missing translation for 'onlineSavingsMsg'
Learn More

Isophthalonitrile 98.0+%, TCI America™

Catalog No. p-7131242
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I015825G 25 g
I0158250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I015825G Supplier TCI America Supplier No. I015825G
Only null left

Chemical Identifiers

CAS 626-17-5
Molecular Formula C8H4N2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00001795
InChI Key LAQPNDIUHRHNCV-UHFFFAOYSA-N
Synonym isophthalonitrile, 1,3-dicyanobenzene, isophthalodinitrile, 1,3-benzenedicarbonitrile, m-dicyanobenzene, 3-cyanobenzonitrile, m-benzenedinitrile, isoftalodinitril, 1,3-benzodinitrile, m-phthalodinitrile
PubChem CID 12276
ChEBI CHEBI:38218
IUPAC Name benzene-1,3-dicarbonitrile
SMILES N#CC1=CC(=CC=C1)C#N

Specifications

Melting Point 162°C
Color White
Quantity 25 g
Formula Weight 128.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isophthalonitrile
EINECSNumber (3)-1799
RTECSNumber CZ1900000
TSCA Yes