missing translation for 'onlineSavingsMsg'
Learn More

Isophthalic Dihydrazide 95.0+%, TCI America™

Catalog No. p-7131296
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I025725G 25 g
I0257100G 100 g
I0257500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I025725G Supplier TCI America Supplier No. I025725G

Chemical Identifiers

CAS 2760-98-7
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.194
MDL Number MFCD00025117
InChI Key UTTHLMXOSUFZCQ-UHFFFAOYSA-N
Synonym isophthalic dihydrazide, isophthalohydrazide, isophthalic acid dihydrazide, isophthalyl dihydrazide, 1,3-benzenedicarboxylic acid, dihydrazide, isophthalic hydrazide, isophthalic acid hydrazide, isophthalic acid, dihydrazide, 1,3-phenylenebiscarbohydrazide, unii-r51j8kl2je
PubChem CID 72700
IUPAC Name benzene-1,3-dicarbohydrazide
SMILES C1=CC(=CC(=C1)C(=O)NN)C(=O)NN

Specifications

Melting Point 227°C
Color White
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isophthalic Dihydrazide
EINECSNumber (3)-1336
TSCA Yes