missing translation for 'onlineSavingsMsg'
Learn More

Isophthalamide 97.0+%, TCI America™

Catalog No. I015225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I015225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I015225G Supplier TCI America Supplier No. I015225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1740-57-4
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
MDL Number MFCD00014801
InChI Key QZUPTXGVPYNUIT-UHFFFAOYSA-N
Synonym isophthalamide, 1,3-benzenedicarboxamide, m-phthalamide, isophthaldiamide, m-carbamoylbenzamide, isophthalic acid diamide, unii-zp57yml58i, zp57yml58i, iso-phthalamide
PubChem CID 74445
ChEBI CHEBI:38801
IUPAC Name benzene-1,3-dicarboxamide
SMILES C1=CC(=CC(=C1)C(=O)N)C(=O)N

Specifications

Color White
Quantity 25 g
Formula Weight 164.16
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Isophthalamide
RTECSNumber CZ2201000
TSCA No