missing translation for 'onlineSavingsMsg'
Learn More

Isophthalamide, 98%

Catalog No. p-7044194
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0513706 5 g
AAL0513714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0513706 Supplier Thermo Scientific Chemicals Supplier No. L0513706

CAS: 1740-57-4 | C8H8N2O2 | 164.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1740-57-4
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
MDL Number MFCD00014801
InChI Key QZUPTXGVPYNUIT-UHFFFAOYSA-N
Synonym isophthalamide, 1,3-benzenedicarboxamide, m-phthalamide, isophthaldiamide, m-carbamoylbenzamide, isophthalic acid diamide, unii-zp57yml58i, zp57yml58i, iso-phthalamide
PubChem CID 74445
ChEBI CHEBI:38801
IUPAC Name benzene-1,3-dicarboxamide
SMILES C1=CC(=CC(=C1)C(=O)N)C(=O)N

Specifications

Melting Point 281°C to 282°C
Quantity 5 g
Beilstein 2045544
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material Isophthalamide
EINECSNumber 217-104-9
RTECSNumber CZ2201000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only