missing translation for 'onlineSavingsMsg'
Learn More

Isophthalaldehydic Acid 98.0+%, TCI America™

Catalog No. I08511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08511G 1 g
I08515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08511G Supplier TCI America Supplier No. I08511G

Chemical Identifiers

CAS 619-21-6
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00039575
InChI Key UHDNUPHSDMOGCR-UHFFFAOYSA-N
Synonym 3-carboxybenzaldehyde, benzoic acid, 3-formyl, m-formyl benzoic acid, 3-formyl benzoic acid, 3-formyl-benzoic acid, unii-hb95grm95d, m-carboxybenzaldehyde, hb95grm95d, 3-formylbenzoicacid, isophthalaldehydsaure
PubChem CID 12077
IUPAC Name 3-formylbenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C=O

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 1 g
Formula Weight 150.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isophthalaldehydic Acid
EINECSNumber (3)-1670
TSCA Yes