missing translation for 'onlineSavingsMsg'
Learn More

Isophoronediamine (cis- and trans- mixture) 99.0+%, TCI America™

Catalog No. I0228500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0228500ML 500 mL
I022825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0228500ML Supplier TCI America Supplier No. I0228500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2855-13-2
Molecular Formula C10H22N2
Molecular Weight (g/mol) 170.30
MDL Number MFCD00019397
InChI Key RNLHGQLZWXBQNY-UHFFFAOYNA-N
Synonym isophorone diamine, isophoronediamine, 3-aminomethyl-3,5,5-trimethylcyclohexylamine, 3-aminomethyl-3,5,5-trimethylcyclohexanamine, cyclohexanemethanamine, 5-amino-1,3,3-trimethyl, ipda, ccris 6680, 5-amino-1,3,3-trimethylcyclohexanemethylamine, cyclohexanemethylamine, 5-amino-1,3,3-trimethyl, 1-amino-3-aminomethyl-3,5,5-trimethylcyclohexane
PubChem CID 17857
IUPAC Name 3-(aminomethyl)-3,5,5-trimethylcyclohexan-1-amine
SMILES CC1(C)CC(N)CC(C)(CN)C1

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 247°C
Quantity 500 mL
UN Number 2289
Formula Weight 170.30
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isophoronediamine (cis- and trans- mixture)
EINECSNumber (3)-2286
TSCA Yes