missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isophorone 97.0+%, TCI America™
$29.83 - $39.14
Chemical Identifiers
| CAS | 78-59-1 |
|---|---|
| Molecular Formula | C9H14O |
| Molecular Weight (g/mol) | 138.21 |
| MDL Number | MFCD00001584 |
| InChI Key | HJOVHMDZYOCNQW-UHFFFAOYSA-N |
| Synonym | isophorone, isoacetophorone, isoforone, isoforon, izoforon, isooctopherone, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, 2-cyclohexen-1-one, 3,5,5-trimethyl, 1,1,3-trimethyl-3-cyclohexene-5-one |
| PubChem CID | 6544 |
| ChEBI | CHEBI:34800 |
| IUPAC Name | 3,5,5-trimethylcyclohex-2-en-1-one |
| SMILES | CC1=CC(=O)CC(C)(C)C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I015125ML
|
TCI America
I015125ML |
25 mL |
Each for $29.83
|
|
|||||
|
I0151500ML
|
TCI America
I0151500ML |
500 mL |
Each for $39.14
|
|
|||||
Chemical Identifiers
| 78-59-1 | |
| 138.21 | |
| HJOVHMDZYOCNQW-UHFFFAOYSA-N | |
| 6544 | |
| 3,5,5-trimethylcyclohex-2-en-1-one |
| C9H14O | |
| MFCD00001584 | |
| isophorone, isoacetophorone, isoforone, isoforon, izoforon, isooctopherone, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, 2-cyclohexen-1-one, 3,5,5-trimethyl, 1,1,3-trimethyl-3-cyclohexene-5-one | |
| CHEBI:34800 | |
| CC1=CC(=O)CC(C)(C)C1 |
Specifications
| 78-59-1 | |
| Yellow | |
| C9H14O | |
| 25 mL | |
| HJOVHMDZYOCNQW-UHFFFAOYSA-N | |
| 3,5,5-trimethylcyclohex-2-en-1-one | |
| 6544 | |
| 138.21 | |
| Liquid |
| -8°C | |
| 215°C | |
| MFCD00001584 | |
| isophorone, isoacetophorone, isoforone, isoforon, izoforon, isooctopherone, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, 2-cyclohexen-1-one, 3,5,5-trimethyl, 1,1,3-trimethyl-3-cyclohexene-5-one | |
| CC1=CC(=O)CC(C)(C)C1 | |
| 138.21 | |
| CHEBI:34800 | |
| ≥97.0% (GC) | |
| Isophorone |
Safety and Handling
EINECSNumber : (3)-2381&(3)-2389
RTECSNumber : GW7700000
TSCA : Yes