missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isophorone diisocyanate, 98+%, mixture of isomers
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.0610g/mL |
| Boiling Point | 153.0°C (10.0 mmHg) |
| Flash Point | 163°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (sum of mixed isomers) (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4830 to 1.4850 (20°C, 589nm) |
| Quantity | 50 mL |
| Specific Gravity | 1.061 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes skin irritation. May cause an allergic skin reaction. May cause respiratory irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Toxic to aquatic life with long lasti |
| Precautionary Statement | GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. If skin irritation or |
| EINECSNumber | 223-861-6 |
RUO – Research Use Only