Learn More
Isophorone, 97%
CAS: 78-59-1 | C9H14O | 138.21 g/mol
$49.07 - $185.36
Chemical Identifiers
| CAS | 78-59-1 |
|---|---|
| Molecular Formula | C9H14O |
| Molecular Weight (g/mol) | 138.21 |
| MDL Number | MFCD00001584 |
| InChI Key | HJOVHMDZYOCNQW-UHFFFAOYSA-N |
| Synonym | isophorone, isoacetophorone, isoforone, isoforon, izoforon, isooctopherone, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, 2-cyclohexen-1-one, 3,5,5-trimethyl, 1,1,3-trimethyl-3-cyclohexene-5-one |
| PubChem CID | 6544 |
| ChEBI | CHEBI:34800 |
| IUPAC Name | 3,5,5-trimethylcyclohex-2-en-1-one |
| SMILES | CC1=CC(=O)CC(C)(C)C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA14844AK
|
Thermo Scientific Chemicals
A14844AK |
250 mL |
Each for $49.07
|
|
|||||
|
AAA14844AP
|
Thermo Scientific Chemicals
A14844AP |
500 mL |
Each for $57.21
|
|
|||||
|
AAA148440F
|
Thermo Scientific Chemicals
A148440F |
2500 mL |
Each for $185.36
|
|
|||||
Description
Isophorone is used as a solvent and as an intermediate in organic synthesis. Isophorone also occurs naturally in cranberries. Isophorone is used as a solvent in some printing inks, paints, lacquers, adhesives, copolymers, coatings, finishings
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsophorone is used as a solvent and as an intermediate in organic synthesis. Isophorone also occurs naturally in cranberries. Isophorone is used as a solvent in some printing inks, paints, lacquers, adhesives, copolymers, coatings, finishings
Solubility
Soluble in water (12g/L).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 78-59-1 | |
| 138.21 | |
| HJOVHMDZYOCNQW-UHFFFAOYSA-N | |
| 6544 | |
| 3,5,5-trimethylcyclohex-2-en-1-one |
| C9H14O | |
| MFCD00001584 | |
| isophorone, isoacetophorone, isoforone, isoforon, izoforon, isooctopherone, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, 2-cyclohexen-1-one, 3,5,5-trimethyl, 1,1,3-trimethyl-3-cyclohexene-5-one | |
| CHEBI:34800 | |
| CC1=CC(=O)CC(C)(C)C1 |
Specifications
| 78-59-1 | |
| 0.922 | |
| 84°C (183°F) | |
| 1.478 | |
| 250 mL | |
| 14,5196 | |
| Soluble in water (12g/L). | |
| CC1=CC(=O)CC(C)(C)C1 | |
| 138.21 | |
| CHEBI:34800 | |
| 97% |
| -8°C | |
| 99°C (18 mmHg) | |
| C9H14O | |
| MFCD00001584 | |
| 1280721 | |
| isophorone, isoacetophorone, isoforone, isoforon, izoforon, isooctopherone, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, 2-cyclohexen-1-one, 3,5,5-trimethyl, 1,1,3-trimethyl-3-cyclohexene-5-one | |
| HJOVHMDZYOCNQW-UHFFFAOYSA-N | |
| 3,5,5-trimethylcyclohex-2-en-1-one | |
| 6544 | |
| 138.21 | |
| Isophorone |
Safety and Handling
GHS H Statement
H351-H302-H312-H319-H335-H227
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P201-P202-P210-P235-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P363-P370+P378q-P501c
H227-H302+H312-H319-H335-H351
EINECSNumber : 201-126-0
RTECSNumber : GW7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only