missing translation for 'onlineSavingsMsg'
Learn More

Isopentylamine, 99%

Catalog No. aal0226014
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0226014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0226014 Supplier Thermo Scientific Chemicals Supplier No. L0226014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-85-7 | C5H13N | 87.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-85-7
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008203
InChI Key BMFVGAAISNGQNM-UHFFFAOYSA-N
Synonym isoamylamine, isopentylamine, 3-methylbutylamine, 1-amino-3-methylbutane, monoisoamylamine, leucamine, 1-butanamine, 3-methyl, 3-methylbutanamine, isovalerylamine, monoisopentylamine
PubChem CID 7894
ChEBI CHEBI:43689
IUPAC Name 3-methylbutan-1-amine
SMILES CC(C)CCN

Specifications

Melting Point -60°C
Density 0.751
pH 12.1
Boiling Point 95°C to 97°C
Flash Point −1°C (30°F)
Odor Amine-like
Refractive Index 1.408
Quantity 25 g
UN Number UN1106
Beilstein 1209230
Sensitivity Air Sensitive
Merck Index 14,5112
Formula Weight 87.17
Percent Purity 99%
Chemical Name or Material Isopentylamine
Show More Show Less
Hazard Category H225-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H225-H301-H314-H318
Highly flammable liquid and vapor.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYLAMINES
EINECSNumber 203-526-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only