Learn More
Isopentyl nitrite, 97%, stab. with 0.2% anhyd. sodium carbonate
CAS: 110-46-3 | C5H11NO2 | 117.15 g/mol
Supplier: Thermo Scientific Chemicals L13909AE
Description
Isopentyl nitrite is used in the conversion of 2,6-disubstituted phenols to diphenoquinones and aminophenols to quinone diazides. It acts as an oxidant and nitrosating agent. It finds application in aromatic arylation, deamination of arylamines using tetrahydrofuran as a hydrogen donor. It is also useful for the preparation of benzyne using anthranilic acid. It is an antihypertensive and vasodilator to treat heart diseases such as angina and an antidote for cyanide poisoning. Further, it is used as a solvent and cleaning agent for a printed circuit board. In addition to this, it is considered to be a replacement for freon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilityMiscible with methanol, chloroform and ether. Slightly miscible with water.
Notes
Air, moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and alcohol.
Specifications
| Isopentyl nitrite | |
| 110-46-3 | |
| 95°C to 100°C | |
| Sweet | |
| 1.387 | |
| 100 mL | |
| 969510 | |
| 14,5123 | |
| Miscible with methanol,chloroform and ether. Slightly miscible with water. | |
| CC(C)CCON=O | |
| 117.15 | |
| CHEBI:2691 | |
| 97% |
| stabilized with 0.2% anhyd. sodium carbonate | |
| 0.875 | |
| −20°C (−4°F) | |
| C5H11NO2 | |
| MFCD00002057 | |
| UN1113 | |
| Air and light sensitive | |
| isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i | |
| OWFXIOWLTKNBAP-UHFFFAOYSA-N | |
| 3-methylbutyl nitrite | |
| 8053 | |
| 117.15 |
Chemical Identifiers
| 110-46-3 | |
| 117.15 | |
| OWFXIOWLTKNBAP-UHFFFAOYSA-N | |
| 8053 | |
| 3-methylbutyl nitrite |
| C5H11NO2 | |
| MFCD00002057 | |
| isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i | |
| CHEBI:2691 | |
| CC(C)CCON=O |
Safety and Handling
GHS H Statement
H225-H331-H302
Highly flammable liquid and vapor.
Toxic if inhaled.
Harmful if swallowed.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P370+P378q-P501c
H225-H302+H332
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL NITRITE
EINECSNumber : 203-770-8
RTECSNumber : NT0187500
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only