missing translation for 'onlineSavingsMsg'
Learn More

Isopentyl nitrite, 97%, stab. with 0.2% anhyd. sodium carbonate

Catalog No. AAL13909AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13909AC 25 mL
AAL13909AE 100 mL
AAL13909AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL13909AC Supplier Thermo Scientific Chemicals Supplier No. L13909AC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 110-46-3 | C5H11NO2 | 117.15 g/mol

Isopentyl nitrite is used in the conversion of 2,6-disubstituted phenols to diphenoquinones and aminophenols to quinone diazides. It acts as an oxidant and nitrosating agent. It finds application in aromatic arylation, deamination of arylamines using tetrahydrofuran as a hydrogen donor. It is also useful for the preparation of benzyne using anthranilic acid. It is an antihypertensive and vasodilator to treat heart diseases such as angina and an antidote for cyanide poisoning. Further, it is used as a solvent and cleaning agent for a printed circuit board. In addition to this, it is considered to be a replacement for freon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with methanol, chloroform and ether. Slightly miscible with water.

Notes
Air, moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and alcohol.

Chemical Identifiers

CAS 110-46-3
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00002057
InChI Key OWFXIOWLTKNBAP-UHFFFAOYSA-N
Synonym isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i
PubChem CID 8053
ChEBI CHEBI:2691
IUPAC Name 3-methylbutyl nitrite
SMILES CC(C)CCON=O

Specifications

Density 0.875
Boiling Point 95°C to 100°C
Flash Point −20°C (−4°F)
Odor Sweet
Refractive Index 1.387
Quantity 25 mL
UN Number UN1113
Beilstein 969510
Sensitivity Air and light sensitive
Merck Index 14,5123
Solubility Information Miscible with methanol,chloroform and ether. Slightly miscible with water.
Formula Weight 117.15
Percent Purity 97%
Chemical Name or Material Isopentyl nitrite, stabilized with 0.2% anhyd. sodium carbonate
Show More Show Less
Hazard Category H225-H302+H332
Hazard Statement GHS H Statement
H225-H331-H302
Highly flammable liquid and vapor.
Toxic if inhaled.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL NITRITE
EINECSNumber 203-770-8
RTECSNumber NT0187500
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only