missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Isonipecotic Acid, 99%

Supplier:  Thermo Scientific™ 122625000

Quantity 500g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC122625000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

498-94-2
129.16
SRJOCJYGOFTFLH-UHFFFAOYSA-N
3773
OC(=O)C1CCNCC1
C6H11NO2
MFCD00006004
isonipecotic acid, 4-piperidinecarboxylic acid, 4-carboxypiperidine, hexahydroisonicotinic acid, dl-isopipecolinic acid, isonicotinic acid, hexahydro, h-inp-oh, acide isonipecotique, 4-hexahydroisonicotinic acid, unii-m5tzp1rwie
piperidine-4-carboxylic acid

Specifications

Isonipecotic acid
1% max. (120°C, 4 hrs)
99%
C6H11NO2
MFCD00006004
15, 5235
(10% in water) Clear to slightly hazy
OC(=O)C1CCNCC1
129.16
129.16
498-94-2
Authentic
Glass bottle
500g
22, V,1, 244
isonipecotic acid, 4-piperidinecarboxylic acid, 4-carboxypiperidine, hexahydroisonicotinic acid, dl-isopipecolinic acid, isonicotinic acid, hexahydro, h-inp-oh, acide isonipecotique, 4-hexahydroisonicotinic acid, unii-m5tzp1rwie
SRJOCJYGOFTFLH-UHFFFAOYSA-N
piperidine-4-carboxylic acid
3773
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-872-3

SDS
Documents

Documents