missing translation for 'onlineSavingsMsg'
Learn More

Isonipecotamide 98.0+%, TCI America™

Catalog No. I025525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I025525G 25 g
I0255250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I025525G Supplier TCI America Supplier No. I025525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39546-32-2
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.18
MDL Number MFCD00038012
InChI Key DPBWFNDFMCCGGJ-UHFFFAOYSA-N
Synonym isonipecotamide, 4-piperidinecarboxamide, hexahydroisonicotinamide, 4-carbamoylpiperidine, isonipectoamide, piperidine-4-carboxylic acid amide, 4-piperdinecarboxamide, piperidin-4-carboxamid, iso nipecotamide, iso-nipecotamide
PubChem CID 3772
IUPAC Name piperidine-4-carboxamide
SMILES NC(=O)C1CCNCC1

Specifications

Melting Point 150°C
Color White
Quantity 25 g
Formula Weight 128.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isonipecotamide