Learn More
Isonipecotamide, 98%
CAS: 39546-32-2 | C6H12N2O | 128.18 g/mol
Supplier: Thermo Scientific Chemicals A1344218
Description
Isonipecotamide is used as a reactant for the preparation of Survival motor neuron (SMN) protein modulators, orally available naphthyridine protein kinase D inhibitors and heteroalicyclic carboxamidines as inhibitors of inducible nitric oxide synthase. It is also used to prepare phosphodiesterase 5 inhibitors. It is also used as a reactant for identification of small molecular inhibitors of importin-beta mediated nuclear import. It is also used as a fluorenylmethyloxycarbonyl - deprotection agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsonipecotamide is used as a reactant for the preparation of Survival motor neuron (SMN) protein modulators, orally available naphthyridine protein kinase D inhibitors and heteroalicyclic carboxamidines as inhibitors of inducible nitric oxide synthase. It is also used to prepare phosphodiesterase 5 inhibitors. It is also used as a reactant for identification of small molecular inhibitors of importin-beta mediated nuclear import. It is also used as a fluorenylmethyloxycarbonyl - deprotection agent in organic synthesis.
Solubility
Soluble in water.
Notes
Moisture sensitive. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
| Isonipecotamide | |
| 148°C to 149°C | |
| MFCD00038012 | |
| 112554 | |
| isonipecotamide, 4-piperidinecarboxamide, hexahydroisonicotinamide, 4-carbamoylpiperidine, isonipectoamide, piperidine-4-carboxylic acid amide, 4-piperdinecarboxamide, piperidin-4-carboxamid, iso nipecotamide, iso-nipecotamide | |
| DPBWFNDFMCCGGJ-UHFFFAOYSA-N | |
| piperidine-4-carboxamide | |
| 3772 | |
| 98% |
| 39546-32-2 | |
| C6H12N2O | |
| 50 g | |
| Hygroscopic | |
| Soluble in water. | |
| NC(=O)C1CCNCC1 | |
| 128.18 | |
| 128.18 |
Chemical Identifiers
| 39546-32-2 | |
| 128.18 | |
| DPBWFNDFMCCGGJ-UHFFFAOYSA-N | |
| 3772 | |
| NC(=O)C1CCNCC1 |
| C6H12N2O | |
| MFCD00038012 | |
| isonipecotamide, 4-piperidinecarboxamide, hexahydroisonicotinamide, 4-carbamoylpiperidine, isonipectoamide, piperidine-4-carboxylic acid amide, 4-piperdinecarboxamide, piperidin-4-carboxamid, iso nipecotamide, iso-nipecotamide | |
| piperidine-4-carboxamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 254-501-6
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only