missing translation for 'onlineSavingsMsg'
Learn More

Isonipecotamide, 98%

Catalog No. AAA1344209
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1344209 10 g
AAA1344218 50 g
AAA1344230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1344209 Supplier Thermo Scientific Chemicals Supplier No. A1344209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 39546-32-2 | C6H12N2O | 128.18 g/mol

Isonipecotamide is used as a reactant for the preparation of Survival motor neuron (SMN) protein modulators, orally available naphthyridine protein kinase D inhibitors and heteroalicyclic carboxamidines as inhibitors of inducible nitric oxide synthase. It is also used to prepare phosphodiesterase 5 inhibitors. It is also used as a reactant for identification of small molecular inhibitors of importin-beta mediated nuclear import. It is also used as a fluorenylmethyloxycarbonyl - deprotection agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isonipecotamide is used as a reactant for the preparation of Survival motor neuron (SMN) protein modulators, orally available naphthyridine protein kinase D inhibitors and heteroalicyclic carboxamidines as inhibitors of inducible nitric oxide synthase. It is also used to prepare phosphodiesterase 5 inhibitors. It is also used as a reactant for identification of small molecular inhibitors of importin-beta mediated nuclear import. It is also used as a fluorenylmethyloxycarbonyl - deprotection agent in organic synthesis.

Solubility
Soluble in water.

Notes
Moisture sensitive. Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 39546-32-2
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.18
MDL Number MFCD00038012
InChI Key DPBWFNDFMCCGGJ-UHFFFAOYSA-N
Synonym isonipecotamide, 4-piperidinecarboxamide, hexahydroisonicotinamide, 4-carbamoylpiperidine, isonipectoamide, piperidine-4-carboxylic acid amide, 4-piperdinecarboxamide, piperidin-4-carboxamid, iso nipecotamide, iso-nipecotamide
PubChem CID 3772
IUPAC Name piperidine-4-carboxamide
SMILES NC(=O)C1CCNCC1

Specifications

Melting Point 148°C to 149°C
Quantity 10 g
Beilstein 112554
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 128.18
Percent Purity 98%
Chemical Name or Material Isonipecotamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 254-501-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only