missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isonicotinoyl Chloride Hydrochloride 95.0+%, TCI America™
$155.93 - $956.80
Chemical Identifiers
| CAS | 39178-35-3 |
|---|---|
| Molecular Formula | C6H5Cl2NO |
| Molecular Weight (g/mol) | 178.01 |
| MDL Number | MFCD00012830 |
| InChI Key | BNTRVUUJBGBGLZ-UHFFFAOYSA-N |
| Synonym | isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl |
| PubChem CID | 12262826 |
| IUPAC Name | hydrogen pyridine-4-carbonyl chloride chloride |
| SMILES | [H+].[Cl-].ClC(=O)C1=CC=NC=C1 |
Chemical Identifiers
| 39178-35-3 | |
| 178.01 | |
| BNTRVUUJBGBGLZ-UHFFFAOYSA-N | |
| 12262826 | |
| [H+].[Cl-].ClC(=O)C1=CC=NC=C1 |
| C6H5Cl2NO | |
| MFCD00012830 | |
| isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl | |
| hydrogen pyridine-4-carbonyl chloride chloride |
Specifications
| 39178-35-3 | |
| White | |
| MFCD00012830 | |
| 3261 | |
| BNTRVUUJBGBGLZ-UHFFFAOYSA-N | |
| hydrogen pyridine-4-carbonyl chloride chloride | |
| 12262826 | |
| ≥95.0% (T) | |
| Isonicotinoyl Chloride Hydrochloride |
| 160°C | |
| C6H5Cl2NO | |
| 25 g | |
| isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl | |
| [H+].[Cl-].ClC(=O)C1=CC=NC=C1 | |
| 178.01 | |
| 178.01 | |
| Crystalline Powder |
Safety and Handling
TSCA : No