missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinoyl chloride hydrochloride, 97%

CAS: 39178-35-3 | C6H5Cl2NO | 178.01 g/mol

$82.81 - $1013.29

Chemical Identifiers

CAS 39178-35-3
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.01
MDL Number MFCD00012830
InChI Key BNTRVUUJBGBGLZ-UHFFFAOYSA-N
Synonym isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl
PubChem CID 12262826
IUPAC Name pyridine-4-carbonyl chloride;hydrochloride
SMILES [H+].[Cl-].ClC(=O)C1=CC=NC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1086309
View Documents
Thermo Scientific Chemicals
A1086309
10 g
Each for $82.81
Only null left
 
AAA1086318
View Documents
Thermo Scientific Chemicals
A1086318
50 g
Each for $263.03
Only null left
 
AAA1086330
View Documents
Thermo Scientific Chemicals
A1086330
250 g
Each for $1,013.29
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39178-35-3
178.01
BNTRVUUJBGBGLZ-UHFFFAOYSA-N
12262826
[H+].[Cl-].ClC(=O)C1=CC=NC=C1
C6H5Cl2NO
MFCD00012830
isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl
pyridine-4-carbonyl chloride;hydrochloride

Specifications

39178-35-3
C6H5Cl2NO
10 g
3626052
isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl
[H+].[Cl-].ClC(=O)C1=CC=NC=C1
178.01
178.01
Isonicotinoyl chloride hydrochloride
154°C to 162°C
MFCD00012830
UN3261
Hygroscopic
BNTRVUUJBGBGLZ-UHFFFAOYSA-N
pyridine-4-carbonyl chloride;hydrochloride
12262826
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 254-331-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only