missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinoyl chloride hydrochloride, 97%

Catalog No. p-3736757
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Packaging:
Plastic bottle
Catalog No. Quantity Packaging
AC211680250 25 g Plastic bottle
1 options

Catalog No. AC211680250

Supplier: Thermo Scientific Chemicals 211680250

Only null left

CAS: 39178-35-3 | C6H5Cl2NO | 178.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39178-35-3
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.01
MDL Number MFCD00012830
InChI Key BNTRVUUJBGBGLZ-UHFFFAOYSA-N
Synonym isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl
PubChem CID 12262826
IUPAC Name pyridine-4-carbonyl chloride;hydrochloride
SMILES [H+].[Cl-].ClC(=O)C1=CC=NC=C1

Specifications

Melting Point 153.0°C to 161.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Plastic bottle
Quantity 25 g
Beilstein 22, 46
Formula Weight 178.02
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Isonicotinoyl chloride hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 254-331-2

RUO – Research Use Only